Silver ≥99.999%, needles

Supplier: BeanTown Chemical

223205-10G 223205-50G
BT223205-10GEA 111 USD
BT223205-10G BT223205-50G
Silver ≥99.999%, needles
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000 UN No: UN3077; Haz Class: 9; Packing Group: III Needles; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212°; Flash point: 111°C (232°F) Density (g/mL): 10.49 Air Sensitive


  • RTECS: VW3500000
  • Air Sensitive
  • Needles

Formula: Ag
Boiling Pt: 2212 °C
Melting Pt: 960 °C
Density: 10.49 g/mL
Flash Pt: 111 °C (232 °F)
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
ADR: 9,III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR