Titanium ≥99.5% (trace metals basis), powder -325 mesh
Supplier: BeanTown Chemical
223200-100G
223200-500G
BT223200-100GEA
79.09
USD
BT223200-100G
BT223200-500G
Titanium ≥99.5% (trace metals basis), powder -325 mesh
Titanium
CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264; RTECS: XR1700000
UN No: UN2546; Haz Class: 4.2; Packing Group: II
Powder; Molecular Formula: Ti; MW: 47.87
Melting Point: 1660°; Boiling Point: 3287°
Density (g/mL): 4.5
- RTECS: XR1700000
- Powder
Formula:
Ti Boiling Pt: 3287 °C Melting Pt: 1660 °C Density: 4.5 g/mL |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 UN: 2546 ADR: 4.2,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...