2-(Dimethylamino)-ethanol ≥99%
Supplier: BeanTown Chemical
CAS: 108-01-0; EC No: 203-542-8; MDL No: MFCD00002846; RTECS: KK6125000
UN No: UN2051; Haz Class: 8 (3); Packing Group: II
Liquid; Molecular Formula: C4H11NO; MW: 89.14
Melting Point: -70°; Boiling Point: 134-136°; Flash point: 39°C (102°F)
Density (g/mL): 0.886; Refractive Index: 1.429
Hygroscopic
- RTECS: KK6125000
- Refractive index: 1.429
- Hygroscopic
- Liquid
Formula:
C₄H₁₁NO Boiling Pt: 134…136 °C Melting Pt: –70 °C Density: 0.886 g/mL Flash Pt: 39 °C (102 °F) |
MDL Number:
MFCD00002846 CAS Number: 108-01-0 EINECS: 203-542-8 UN: 2051 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...