Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane ≥97%

Supplier: BeanTown Chemical

Synonyms: 1H,1H,2H,2H-Perfluorodecyltriethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane, Triethoxy-1H,1H,2H,2H-perfluorodecylsilane

223050-1G 223050-25G 223050-5G
BT223050-1GEA 36 USD
BT223050-1G BT223050-25G BT223050-5G
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane ≥97%
Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane

CAS: 101947-16-4; MDL No: MFCD00042334 UN No: UN1760; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C16F17H19O3Si; MW: 610.38 Boiling Point: 209-230°; Flash point: 110°C (230°F) Density (g/mL): 1.389; Refractive Index: 1.342 Moisture Sensitive


  • Refractive index: 1.342
  • Moisture Sensitive
  • Liquid

Formula: C₁₆H₁₉F₁₇O₃Si
MW: 610.38 g/mol
Boiling Pt: 209…230 °C
Density: 1.389 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00042334
CAS Number: 101947-16-4
ADR: 8 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR