Dipotassium hydrogen orthophosphate trihydrate
Supplier: BeanTown Chemical
Synonyms:
Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate
222860-1KG
222860-250G
BT222860-1KGEA
172
USD
BT222860-1KG
BT222860-250G
Dipotassium hydrogen orthophosphate trihydrate
di-Potassium hydrogen orthophosphate trihydrate
CAS: 16788-57-1; EC No: 231-834-5; MDL No: MFCD00149926
Crystalline/Powder; Linear Formula: K2HPO4·3H2O; MW: 228.23
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
K₂HPO₄·3H₂O MW: 228.22 g/mol |
MDL Number:
MFCD00149926 CAS Number: 16788-57-1 EINECS: 231-834-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...