di-Sodium tetraborate decahydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
222805-10KG
222805-2.5KG
222805-500G
BT222805-10KGEA
244
USD
BT222805-10KG
BT222805-2.5KG
BT222805-500G
di-Sodium tetraborate decahydrate ≥99%
di-Sodium tetraborate decahydrate
CAS: 1303-96-4; EC No: 215-540-4; MDL No: MFCD00149193; RTECS: VZ2275000
Crystalline/Powder; Linear Formula: Na2B4O7·10H2O; MW: 381.37
Melting Point: 62°
Density (g/mL): 1.73
- RTECS: VZ2275000
- Crystalline/Powder
Formula:
Na₂B₄O₇·10H₂O Melting Pt: 62 °C Density: 1.73 g/mL |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 EINECS: 215-540-4 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...