di-Sodium tetraborate decahydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate

222805-10KG 222805-2.5KG 222805-500G
BT222805-10KGEA 244 USD
BT222805-10KG BT222805-2.5KG BT222805-500G
di-Sodium tetraborate decahydrate ≥99%
di-Sodium tetraborate decahydrate

CAS: 1303-96-4; EC No: 215-540-4; MDL No: MFCD00149193; RTECS: VZ2275000 Crystalline/Powder; Linear Formula: Na2B4O7·10H2O; MW: 381.37 Melting Point: 62° Density (g/mL): 1.73


  • RTECS: VZ2275000
  • Crystalline/Powder

Formula: Na₂B₄O₇·10H₂O
Melting Pt: 62 °C
Density: 1.73 g/mL
MDL Number: MFCD00149193
CAS Number: 1303-96-4
EINECS: 215-540-4

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR