Aluminium oxide, γ-phase ≥99%
Supplier: BeanTown Chemical
222795-100G
222795-25G
222795-500G
BT222795-100GEA
169
USD
BT222795-100G
BT222795-25G
BT222795-500G
Aluminium oxide, γ-phase ≥99%
Aluminium oxide, γ-phase
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
40-80nm APS Powder, S. A. 180 m2/g; Molecular Formula: Al2O3 ; MW: 101.96
Melting Point: 2040°
- 40-80nm APS Powder, S. A. 180 m²/g
Formula:
Al₂O₃ Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...