4-tert-Butylbenzenesulfonyl chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-tert-Butylbenzene-1-sulfonyl chloride, 4-tert-Butylbenzene-1-sulphonyl chloride
222715-10G
222715-50G
BT222715-10GEA
48
USD
BT222715-10G
BT222715-50G
4-tert-Butylbenzenesulfonyl chloride ≥98%
4-tert-Butylbenzenesulphonyl chloride
CAS: 15084-51-2; MDL No: MFCD00007449
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: C10H13ClO2S; MW: 232.73
Melting Point: 78-81°
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
C₁₀H₁₃ClO₂S MW: 232.73 Boiling Pt: 165 Melting Pt: 83 |
MDL Number:
MFCD00007449 CAS Number: 15084-51-2 UN: 3261 ADR: 8 II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...