Lithium hydroxide monohydrate ≥98%
Supplier: BeanTown Chemical
222710-100G
222710-2.5KG
222710-500G
BT222710-100GEA
81.58
USD
BT222710-100G
BT222710-2.5KG
BT222710-500G
Lithium hydroxide monohydrate ≥98%
Lithium hydroxide monohydrate
CAS: 1310-66-3; EC No: 215-183-4; MDL No: MFCD00149772; RTECS: OJ6307080
UN No: UN2680; Haz Class: 8; Packing Group: II
Powder; Linear Formula: LiOH·H2O; MW: 41.96
Density (g/mL): 1.51
Air Sensitive , Hygroscopic
- RTECS: OJ6307080
- Air Sensitive, Hygroscopic
- Powder
Formula:
LiOH·1H₂O Density: 1.51 g/mL |
MDL Number:
MFCD00149772 CAS Number: 1310-66-3 EINECS: 215-183-4 UN: 2680 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...