Disodium-1,6-naphthalenedisulfonate

Supplier: BeanTown Chemical

Synonyms: 1,6-Naphthalenedisulphonic acid Disodium salt, 1,6-Naphthalenedisulfonic acid Disodium salt, Sodium naphthalene-1,6-disulphonate, Sodium naphthalene-1,6-disulfonate

222675-1G
BT222675-1GEA 295 USD
BT222675-1G
Disodium-1,6-naphthalenedisulfonate
Disodium-1,6-naphthalenedisulfonate

CAS: 1655-43-2; EC No: 216-734-1; MDL No: MFCD00021522 Solid; Molecular Formula: C10H6Na2O6S2; MW: 332.26


  • Solid

Formula: C₁₀H₈O₆S₂
MW: 332.25 g/mol
MDL Number: MFCD00021522
CAS Number: 1655-43-2
EINECS: 216-734-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR