tert-Butyl carbamate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Carbamic acid tert-butyl ester
222670-1G
222670-25G
222670-5G
BT222670-1GEA
21
USD
BT222670-1G
BT222670-25G
BT222670-5G
tert-Butyl carbamate ≥98%
tert-Butyl carbamate
CAS: 4248-19-5; EC No: 224-209-3; MDL No: MFCD00007962
Crystalline; Linear Formula: (CH3)3COCONH2; Molecular Formula: C5H11NO2; MW: 117.15
Melting Point: 105-108°
- Crystalline
Formula:
C₅H₁₁NO₂ Melting Pt: 105…108 °C |
MDL Number:
MFCD00007962 CAS Number: 4248-19-5 EINECS: 224-209-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...