Zirconium ≥99.5% (trace metals basis excluding Hf), slug, Length 12.7 mm, Diameter 6.35 mm

Supplier: BeanTown Chemical

222650-100G 222650-25G
BT222650-100GEA 130 USD
BT222650-100G BT222650-25G
Zirconium ≥99.5% (trace metals basis excluding Hf), slug, Length 12.7 mm, Diameter 6.35 mm
Zirconium

CAS: 7440-67-7; EC No: 231-176-9; MDL No: MFCD00011303 Solid; Molecular Formula: Zr; MW: 91.22 Melting Point: 1852°; Boiling Point: 4377° Density (g/mL): 6.5


  • Solid

Formula: Zr
Boiling Pt: 4377 °C
Melting Pt: 1852 °C
Density: 6.5 g/mL
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR