Pyridine ≥99%
Supplier: BeanTown Chemical
Synonyms:
Azabenzene, Azine
222645-10L
222645-2.5L
222645-500ML
BT222645-10LEA
581.46
USD
BT222645-10L
BT222645-2.5L
BT222645-500ML
Pyridine ≥99%
Pyridine
CAS: 110-86-1; EC No: 203-809-9; MDL No: MFCD00011732; RTECS: UR8400000
UN No: UN1282; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C5H5N; MW: 79.10
Melting Point: -42°; Boiling Point: 115°; Flash point: 17°C (62.6°F)
Density (g/mL): 0.978; Refractive Index: 1.509
- RTECS: UR8400000
- Refractive index: 1.509
- Liquid
Formula:
C₅H₅N Boiling Pt: 115 °C Melting Pt: –42 °C Density: 0.978 g/mL Flash Pt: 17 °C (62.6 °F) |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...