N-Carbobenzyloxy-D-proline ≥98%

Supplier: BeanTown Chemical

Synonyms: (R)-1-((Benzyloxy)carbonyl)pyrrolidine-2-carboxylic acid, Z-D-Pro-OH, N-Cbz-D-proline, Cbz-D-Pro-OH, ((Benzyloxy)carbonyl)-D-proline, N-Benzyloxycarbonyl-D-proline

222580-1G 222580-25G 222580-5G
BT222580-1GEA 26 USD
BT222580-1G BT222580-25G BT222580-5G
N-Carbobenzyloxy-D-proline ≥98%
N-Carbobenzyloxy-D-proline

CAS: 6404-31-5; EC No: 229-021-5; MDL No: MFCD00063228 Solid; Molecular Formula: C13H15NO4 ; MW: 249.26 Melting Point: 76-78° Optical Rotation: [α]25/D +40.2°, c = 2 in ethanol


  • Optical rotation: [α]25/D +40.2°, c = 2 in ethanol
  • Solid

Formula: C₁₃H₁₅NO₄
MW: 249.3 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD00063228
CAS Number: 6404-31-5
EINECS: 229-021-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR