Samarium cobalt

Supplier: BeanTown Chemical

222575-250G 222575-50G
BT222575-250GEA 325 USD
BT222575-250G BT222575-50G
Samarium cobalt
Samarium cobalt

CAS: 12017-68-4; EC No: 234-625-7; MDL No: MFCD02091682 UN No: UN3089; Haz Class: 4.1; Packing Group: III Solid, Intermetallic, Magnet Alloy Additive; Linear Formula: SmCo5; MW: 445.03 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • Solid, Intermetallic, Magnet Alloy Additive

Formula: CoSm
MDL Number: MFCD02091682
CAS Number: 12017-68-4
EINECS: 234-625-7
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR