Boron carbide ≥90%, Technical Grade

Supplier: BeanTown Chemical

222540-100G 222540-500G
BT222540-100GEA 48 USD
BT222540-100G BT222540-500G
Boron carbide ≥90%, Technical Grade
Boron carbide

CAS: 12069-32-8; EC No: 235-111-5; MDL No: MFCD00011520; RTECS: ED7420000 22-59 Micron Powder ; Molecular Formula: CB4; MW: 55.25 Density (g/mL): 2.51


  • RTECS: ED7420000
  • 22-59 Micron Powder

Formula: CB₄
MW: 55.25
Melting Pt: 2350
Density: 2.51 g/mL
MDL Number: MFCD00011520
CAS Number: 12069-32-8
EINECS: 235-111-5
Merck Index: 12,01374

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR