Nickel(II) bromide, anhydrous ≥99%
Supplier: BeanTown Chemical
Synonyms:
Nickel dibromide
222455-250G
222455-50G
BT222455-250GEA
466
USD
BT222455-250G
BT222455-50G
Nickel(II) bromide, anhydrous ≥99%
Nickel (II) bromide
CAS: 13462-88-9; EC No: 236-665-0; MDL No: MFCD00011141
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline/Powder; Linear Formula: NiBr2; MW: 218.53
Melting Point: 963°
Density (g/mL): 5.098
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
Br₂Ni Melting Pt: 963 °C Density: 5.098 g/mL |
MDL Number:
MFCD00011141 CAS Number: 13462-88-9 EINECS: 236-665-0 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...