7-Isoquinolinol ≥97%
Supplier: BeanTown Chemical
Synonyms:
7-Hydroxyisoquinoline, Isoquinolin-7-ol
222425-1G
222425-250MG
BT222425-1GEA
161
USD
BT222425-1G
BT222425-250MG
7-Isoquinolinol ≥97%
7-Isoquinolinol
CAS: 7651-83-4; MDL No: MFCD00456131
Powder; Molecular Formula: C9H7NO; MW: 145.16
Melting Point: 226-228°
- Powder
Formula:
C₉H₇NO Melting Pt: 226…228 °C |
MDL Number:
MFCD00456131 CAS Number: 7651-83-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...