Lithium hydroxide, anhydrous ≥99.995%
Supplier: BeanTown Chemical
222390-100G
222390-25G
222390-5G
BT222390-100GEA
513.54
USD
BT222390-100G
BT222390-25G
BT222390-5G
Lithium hydroxide, anhydrous ≥99.995%
Lithium hydroxide
CAS: 1310-65-2; EC No: 215-183-4; MDL No: MFCD00011095; RTECS: OJ6307070
UN No: UN2680; Haz Class: 8; Packing Group: II
Powder; Linear Formula: LiOH; MW: 23.95
Melting Point: 470° (decomposes)
Density (g/mL): 2.54
Air Sensitive , Hygroscopic
- RTECS: OJ6307070
- Air Sensitive, Hygroscopic
- Powder
Formula:
LiOH Melting Pt: 470 °C (dec.) Density: 2.54 g/mL |
MDL Number:
MFCD00011095 CAS Number: 1310-65-2 EINECS: 215-183-4 UN: 2680 ADR: 8,II |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...