Sodium molybdate(VI) dihydrate ≥98%
Supplier: BeanTown Chemical
222330-100G
222330-2.5KG
222330-500G
BT222330-100GEA
49.83
USD
BT222330-100G
BT222330-2.5KG
BT222330-500G
Sodium molybdate(VI) dihydrate ≥98%
Sodium molybdate(VI) dihydrate
CAS: 10102-40-6; EC No: 231-551-7; MDL No: MFCD00149170; RTECS: QA5085000
Crystalline/Powder; Linear Formula: Na2MoO4·2H2O; MW: 241.95
Melting Point: 100° (decomposes)
Density (g/mL): 2.37
- RTECS: QA5085000
- Crystalline/Powder
Formula:
Na₂MoO₄·2H₂O Melting Pt: 100 °C (dec.) Density: 2.37 g/mL |
MDL Number:
MFCD00149170 CAS Number: 10102-40-6 EINECS: 231-551-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...