Potassium perrhenate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Potassium perrhenate (VII)
222325-10G
222325-2G
222325-500MG
BT222325-10GEA
268
USD
BT222325-10G
BT222325-2G
BT222325-500MG
Potassium perrhenate ≥99%
Potassium perrhenate
CAS: 10466-65-6; EC No: 233-953-8; MDL No: MFCD00011365; RTECS: SE1400000
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Crystalline/Powder; Linear Formula: KReO4; MW: 289.30
Melting Point: 550°; Boiling Point: 1360-1370°
Density (g/mL): 4.887; Refractive Index: 1.643
- RTECS: SE1400000
- Refractive index: 1.643
- Crystalline/Powder
Formula:
KO₄Re Boiling Pt: 1360…1370 °C Melting Pt: 550 °C Density: 4.887 g/mL |
MDL Number:
MFCD00011365 CAS Number: 10466-65-6 EINECS: 233-953-8 UN: 1479 ADR: 5.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...