Europium(III) fluoride, anhydrous ≥99.5%

Supplier: BeanTown Chemical

222230-1G 222230-25G 222230-5G
BT222230-1GEA 43 USD
BT222230-1G BT222230-25G BT222230-5G
Europium(III) fluoride, anhydrous ≥99.5%
Europium (III) fluoride

CAS: 13765-25-8; EC No: 237-368-9; MDL No: MFCD00010998 -60 Mesh Powder; Molecular Formula: EuF3; MW: 208.96 Boiling Point: 2280° Hygroscopic


  • Hygroscopic
  • -60 Mesh Powder

Formula: EuF₃
Boiling Pt: 2280 °C
MDL Number: MFCD00010998
CAS Number: 13765-25-8
EINECS: 237-368-9

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR