Acetonedimethyl acetal ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane

222175-100ML 222175-500ML
BT222175-100MLEA 29.57 USD
BT222175-100ML BT222175-500ML
Acetonedimethyl acetal ≥98%
Acetone-dimethyl acetal

CAS: 77-76-9; EC No: 201-056-0; MDL No: MFCD00008479 UN No: UN3271; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3O)2C(CH3)2; Molecular Formula: C5H12O2; MW: 104.15 Boiling Point: 83°; Flash point: -10°C (14°F) Density (g/mL): 0.847; Refractive Index: 1.378


  • Refractive index: 1.378
  • Liquid

Formula: C₅H₁₂O₂
Boiling Pt: 83 °C
Density: 0.847 g/mL
Flash Pt: –10 °C (14 °F)
MDL Number: MFCD00008479
CAS Number: 77-76-9
EINECS: 201-056-0
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR