2-Aminopyridin-4-ol ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Amino-4-hydroxypyridine
222075-1G
222075-5G
BT222075-1GEA
35
USD
BT222075-1G
BT222075-5G
2-Aminopyridin-4-ol ≥98%
2-Aminopyridin-4-ol
CAS: 33631-05-9; MDL No: MFCD04114161
Liquid; Molecular Formula: C5H6N2O ; MW: 110.11
- Liquid
Formula:
C₅H₆N₂O MW: 110.12 g/mol |
MDL Number:
MFCD04114161 CAS Number: 33631-05-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...