2-Aminopyridin-4-ol ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Amino-4-hydroxypyridine

222075-1G 222075-5G
BT222075-1GEA 35 USD
BT222075-1G BT222075-5G
2-Aminopyridin-4-ol ≥98%
2-Aminopyridin-4-ol

CAS: 33631-05-9; MDL No: MFCD04114161 Liquid; Molecular Formula: C5H6N2O ; MW: 110.11


  • Liquid

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD04114161
CAS Number: 33631-05-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR