3-(3,4-Dihydroxyphenyl)-L-alanine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa
222035-100G
222035-25G
222035-5G
BT222035-100GEA
384
USD
BT222035-100G
BT222035-25G
BT222035-5G
3-(3,4-Dihydroxyphenyl)-L-alanine ≥98%
3-(3,4-Dihydroxyphenyl)-L-alanine
CAS: 59-92-7; EC No: 200-445-2; MDL No: MFCD00002598; RTECS: AY5600000
Crystalline; Molecular Formula: C9H11NO4; MW: 197.19
Melting Point: 276-278°
- RTECS: AY5600000
- Crystalline
Formula:
C₉H₁₁NO₄ MW: 197.19 g/mol Melting Pt: 276…278 °C |
MDL Number:
MFCD00002598 CAS Number: 59-92-7 EINECS: 200-445-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...