Iron(III) oxide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide
221995-100G
221995-25G
221995-500G
BT221995-100GEA
235
USD
BT221995-100G
BT221995-25G
BT221995-500G
Iron(III) oxide ≥99.99%
Iron (III) oxide
CAS: 1309-37-1; EC No: 215-168-2; MDL No: MFCD00011008
Powder; Molecular Formula: Fe2O3 ; MW: 159.69
- Powder
Formula:
Fe₂O₃ |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...