Platinum, wire Ø 0.508 mm for ISA type R or S standard grade thermocouple

Supplier: BeanTown Chemical

Synonyms: Platinium

221925-100CM 221925-25CM
BT221925-100CMEA 1016 USD
BT221925-100CM BT221925-25CM
Platinum, wire Ø 0.508 mm for ISA type R or S standard grade thermocouple
Platinum

CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000 Solid; Molecular Formula: Pt; MW: 195.08 Melting Point: 1772°; Boiling Point: 3827° Density (g/mL): 21.45


  • RTECS: TP2160000
  • Solid

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45 g/mL
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR