Antimony(III) telluride ≥99.999%

Supplier: BeanTown Chemical

221890-10G 221890-50G
BT221890-10GEA 116 USD
BT221890-10G BT221890-50G
Antimony(III) telluride ≥99.999%
Antimony (III) telluride

CAS: 1327-50-0; EC No: 215-480-9; MDL No: MFCD00016320 UN No: UN1549; Haz Class: 6.1; Packing Group: III 3-12mm Lump ; Molecular Formula: Sb2Te3; MW: 626.32 Density (g/mL): 6.5


  • 3-12mm Lump

Formula: Sb₂Te₃
Density: 6.5 g/mL
MDL Number: MFCD00016320
CAS Number: 1327-50-0
EINECS: 215-480-9
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR