Lanthanum(III) nitrate hexahydrate ≥99.99%
Supplier: BeanTown Chemical
221860-250G
221860-50G
BT221860-250GEA
208
USD
BT221860-250G
BT221860-50G
Lanthanum(III) nitrate hexahydrate ≥99.99%
Lanthanum (III) nitrate hexahydrate
CAS: 10277-43-7; EC No: 233-238-0; MDL No: MFCD00149751; RTECS: OE5075000
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline Aggregates; Linear Formula: La(NO3)3·6H2O; MW: 433.01
Melting Point: 65-68°
- RTECS: OE5075000
- Crystalline Aggregates
Formula:
La(NO₃)₃·6H₂O Melting Pt: 65…68 °C |
MDL Number:
MFCD00149751 CAS Number: 10277-43-7 EINECS: 233-238-0 UN: 1477 ADR: 5.1,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...