Potassium hexachloroplatinate(IV)

Supplier: BeanTown Chemical

Synonyms: Dipotassium hexachloroplatinate

221855-1G 221855-250MG 221855-5G
BT221855-1GEA 128 USD
BT221855-1G BT221855-250MG BT221855-5G
Potassium hexachloroplatinate(IV)
Potassium hexachloroplatinate(IV)

CAS: 16921-30-5; EC No: 240-979-3; MDL No: MFCD00011389; RTECS: TP1650000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: K2PtCl6; MW: 485.99 Melting Point: 250° (decomposes) Density (g/mL): 3.5


  • RTECS: TP1650000
  • Crystalline/Powder

Formula: K₂PtCl₆
Melting Pt: 250 °C (dec.)
Density: 3.5 g/mL
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR