Silicon(II) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Silicium (II) oxide, Silicon monoxide, Silicon oxide

221830-100G 221830-25G
BT221830-100GEA 397 USD
BT221830-100G BT221830-25G
Silicon(II) oxide ≥99.9%
Silicon (II) oxide

CAS: 10097-28-6; EC No: 233-232-8; MDL No: MFCD00151536 3-6mm Lump, Outgassed; Linear Formula: SiO; MW: 44.09 Melting Point: 1702°; Boiling Point: 1880° Density (g/mL): 2.13


  • 3-6mm Lump, Outgassed

Formula: SiO
MW: 44.08 g/mol
Boiling Pt: 1880
Melting Pt: 1702
Density: 2.13 g/mL
MDL Number: MFCD00151536
CAS Number: 10097-28-6
EINECS: 233-232-8
Merck Index: 12,08640

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR