Sodium hydrogen sulfate monohydrate
Supplier: BeanTown Chemical
CAS: 10034-88-5; EC No: 231-665-7; MDL No: MFCD00149210; RTECS: VZ1860000
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: NaHSO4·H2O; MW: 138.07
Melting Point: 183° (decomposes)
Density (g/mL): 2.103
Moisture Sensitive
- RTECS: VZ1860000
- Moisture Sensitive
- Crystalline
Formula:
NaHSO₄·H₂O Melting Pt: 183 °C (dec.) Density: 2.103 g/mL |
MDL Number:
MFCD00149210 CAS Number: 10034-88-5 EINECS: 231-665-7 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...