DL-Proline ≥99%

Supplier: BeanTown Chemical

Synonyms: (±)-Proline, H-DL-Pro-OH

221740-100G 221740-25G 221740-5G
BT221740-100GEA 637 USD
BT221740-100G BT221740-25G BT221740-5G
DL-Proline ≥99%
DL-Proline

CAS: 609-36-9; EC No: 210-189-3; MDL No: MFCD00005250 Solid; Molecular Formula: C5H9NO2; MW: 115.13 Melting Point: 208° (decomposes)


  • Solid

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 245
Melting Pt: 208 °C (dec.)
MDL Number: MFCD00005250
CAS Number: 609-36-9
EINECS: 210-189-3
Merck Index: 12,07963

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR