Dilithium zirconium trioxide ≥94%

Supplier: BeanTown Chemical

Synonyms: Lithium zirconate, Lithium zirconium oxide

221725-100G 221725-25G 221725-500G
BT221725-100GEA 248.43 USD
BT221725-100G BT221725-25G BT221725-500G
Dilithium zirconium trioxide ≥94%
di-Lithium zirconium trioxide

CAS: 12031-83-3; EC No: 234-760-1; MDL No: MFCD00016182 -80 Mesh Powder; Linear Formula: Li2ZrO3; MW: 153.1


  • -80 Mesh Powder

Formula: Li₂O₃Zr
MDL Number: MFCD00016182
CAS Number: 12031-83-3
EINECS: 234-760-1

Order Now

Specification Test Results

Assay 94%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR