2,4,6-Tri-tert-butylaniline ≥95%

Supplier: BeanTown Chemical

Synonyms: 2,4,6-Tris(tert-butyl)aniline

221685-1G 221685-5G
BT221685-1GEA 50 USD
BT221685-1G BT221685-5G
2,4,6-Tri-tert-butylaniline ≥95%
2,4,6-Tri-tert-butylaniline

CAS: 961-38-6; EC No: 213-507-9; MDL No: MFCD00011645 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C18H31N; MW: 261.45 Melting Point: 145-147° Light Sensitive


  • Light Sensitive
  • Crystalline

Formula: C₁₈H₃₁N
MW: 261.45 g/mol
Boiling Pt: 302
Melting Pt: 145
MDL Number: MFCD00011645
CAS Number: 961-38-6
EINECS: 213-507-9
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR