Cadmium(II) telluride ≥99.999%

Supplier: BeanTown Chemical

Synonyms: Cadmium telluride

221650-100G 221650-25G 221650-5G
BT221650-100GEA 490 USD
BT221650-100G BT221650-25G BT221650-5G
Cadmium(II) telluride ≥99.999%
Cadmium (II) telluride

CAS: 1306-25-8; EC No: 215-149-9; MDL No: MFCD00015998; RTECS: EV3330000 UN No: UN3077; Haz Class: 9; Packing Group: III Polycrystalline Lump; Molecular Formula: CdTe; MW: 240.01 Melting Point: 1092°; Boiling Point: 1130° Density (g/mL): 6.2


  • RTECS: EV3330000
  • Polycrystalline Lump

Formula: CdTe
MW: 240.01 g/mol
Boiling Pt: 1130 °C
Melting Pt: 1089.85
Density: 6.2 g/mL
MDL Number: MFCD00015998
CAS Number: 1306-25-8
EINECS: 215-149-9
UN: 3077
ADR: 6.1 III
Merck Index: 12,01667

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR