Cadmium(II) telluride ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Cadmium telluride
221650-100G
221650-25G
221650-5G
BT221650-100GEA
490
USD
BT221650-100G
BT221650-25G
BT221650-5G
Cadmium(II) telluride ≥99.999%
Cadmium (II) telluride
CAS: 1306-25-8; EC No: 215-149-9; MDL No: MFCD00015998; RTECS: EV3330000
UN No: UN3077; Haz Class: 9; Packing Group: III
Polycrystalline Lump; Molecular Formula: CdTe; MW: 240.01
Melting Point: 1092°; Boiling Point: 1130°
Density (g/mL): 6.2
- RTECS: EV3330000
- Polycrystalline Lump
Formula:
CdTe Boiling Pt: 1130 °C Melting Pt: 1092 °C Density: 6.2 g/mL |
MDL Number:
MFCD00015998 CAS Number: 1306-25-8 EINECS: 215-149-9 UN: 3077 ADR: 6.1,III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...