Cerium(IV) oxide 20% (w/w) in water, colloidal dispersion

Supplier: BeanTown Chemical

Synonyms: Ceric oxide, Cerium dioxide

221580-100G 221580-2KG 221580-500G
BT221580-100GEA 34 USD
BT221580-100G BT221580-2KG BT221580-500G
Cerium(IV) oxide 20% (w/w) in water, colloidal dispersion
Cerium (IV) oxide

CAS: 1306-38-3; EC No: 215-150-4; MDL No: MFCD00010933 0.01-0.02 Micron Particles, in Liquid, CeO2 mol/mol 0.4 acetate, pH 3.0; Molecular Formula: CeO2; MW: 172.12 Boiling Point: 100 Density (g/mL): 1.22


  • 0.01-0.02 Micron Particles, in Liquid, CeO2 mol/mol 0.4 acetate, pH 3.0

Formula: CeO₂
Boiling Pt: 100 °C
Density: 1.22 g/mL
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR