Iridium ≥99.95% (trace metals basis), Powder -60 mesh

Supplier: BeanTown Chemical

221560-10G 221560-2G 221560-500MG
BT221560-10GEA 4144 USD
BT221560-10G BT221560-2G BT221560-500MG
Iridium ≥99.95% (trace metals basis), Powder -60 mesh
Iridium

CAS: 7439-88-5; EC No: 231-095-9; MDL No: MFCD00011062 Powder; Molecular Formula: Ir; MW: 192.22 Melting Point: 2450°; Boiling Point: 4130° Density (g/mL): 22.65


  • Powder

Formula: Ir
Boiling Pt: 4130 °C
Melting Pt: 2450 °C
Density: 22.65 g/mL
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR