2-Methoxy-3-nitropyridine ≥98%
Supplier: BeanTown Chemical
221550-1G
221550-25G
221550-5G
BT221550-1GEA
37
USD
BT221550-1G
BT221550-25G
BT221550-5G
2-Methoxy-3-nitropyridine ≥98%
2-Methoxy-3-nitropyridine
CAS: 20265-35-4; MDL No: MFCD00023459
Solid; Molecular Formula: C6H6N2O3; MW: 154.12
Melting Point: 52-57°
- Solid
Formula:
C₆H₆N₂O₃ Melting Pt: 52…57 °C |
MDL Number:
MFCD00023459 CAS Number: 20265-35-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...