1,2-Epoxycyclopentane ≥97%
Supplier: BeanTown Chemical
CAS: 285-67-6; EC No: 206-005-6; MDL No: MFCD00005161; RTECS: RN8935000
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C5H8O; MW: 84.12
Boiling Point: 102°; Flash point: 10°C (50°F)
Density (g/mL): 0.964; Refractive Index: 1.434
- RTECS: RN8935000
- Refractive index: 1.434
- Liquid
Formula:
C₅H₈O Boiling Pt: 102 °C Density: 0.964 g/mL Flash Pt: 10 °C (50 °F) |
MDL Number:
MFCD00005161 CAS Number: 285-67-6 EINECS: 206-005-6 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...