Lithium sulfate, anhydrous ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
221470-100G
221470-25G
221470-500G
BT221470-100GEA
412
USD
BT221470-100G
BT221470-25G
BT221470-500G
Lithium sulfate, anhydrous ≥99.99% (trace metals basis)
Lithium sulphate
CAS: 10377-48-7; EC No: 233-820-4; MDL No: MFCD00011086
Powder; Linear Formula: Li2SO4; MW: 109.94
Melting Point: 845°
Density (g/mL): 2.22
Hygroscopic
- Hygroscopic
- Powder
Formula:
Li₂O₄S Melting Pt: 845 °C Density: 2.22 g/mL |
MDL Number:
MFCD00011086 CAS Number: 10377-48-7 EINECS: 233-820-4 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...