Malononitrile ≥99%
Supplier: BeanTown Chemical
CAS: 109-77-3; EC No: 203-703-2; MDL No: MFCD00001883; RTECS: OO3150000
UN No: UN2647; Haz Class: 6.1; Packing Group: II
Solid; Linear Formula: CH2(CHN)2; MW: 66.06
Melting Point: 30-32°; Boiling Point: 220°; Flash point: 86°C (187°F)
Density (g/mL): 1.049; Refractive Index: 1.415
- RTECS: OO3150000
- Refractive index: 1.415
- Solid
Formula:
C₃H₂N₂ Boiling Pt: 220 °C Melting Pt: 30…32 °C Density: 1.049 g/mL Flash Pt: 86 °C (187 °F) |
MDL Number:
MFCD00001883 CAS Number: 109-77-3 EINECS: 203-703-2 UN: 2647 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...