1,4-Butanediol ≥99%
Supplier: BeanTown Chemical
CAS: 110-63-4; EC No: 203-786-5; MDL No: MFCD00002968; RTECS: EK0525000
Liquid; Linear Formula: HO(CH2)4OH; Molecular Formula: C4H10O2; MW: 90.12
Melting Point: 16°; Boiling Point: 230°; Flash point: 135°C (275°F)
Density (g/mL): 1.017; Refractive Index: 1.445
Hygroscopic
- RTECS: EK0525000
- Refractive index: 1.445
- Hygroscopic
- Liquid
Formula:
C₄H₁₀O₂ Boiling Pt: 230 °C Melting Pt: 16 °C Density: 1.017 g/mL Flash Pt: 135 °C (275 °F) |
MDL Number:
MFCD00002968 CAS Number: 110-63-4 EINECS: 203-786-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...