6-Chlorouracil ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Chloro-2,4-dihydroxypyrimidine, 6-Chloropyrimidine-2,4(1H,3H)-dione, 4-Chlorouracil, 4-(3-Bromophenyl)pyrazole, 4-Chloro-2,6-dihydroxypyrimidine, 6-Chloro-2,4(1H,3H)-pyrimidinedione

221385-1G 221385-5G
BT221385-1GEA 52 USD
BT221385-1G BT221385-5G
6-Chlorouracil ≥98%
6-Chlorouracil

CAS: 4270-27-3; EC No: 224-258-0; MDL No: MFCD00014595 Solid; Molecular Formula: C4H3ClN2O2; MW: 146.53


  • Solid

Formula: C₄H₃ClN₂O₂
MDL Number: MFCD00014595
CAS Number: 4270-27-3
EINECS: 224-258-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR