Trimethyl orthopropionate ≥95%
Supplier: BeanTown Chemical
221345-25ML
BT221345-25MLEA
148
USD
BT221345-25ML
Trimethyl orthopropionate ≥95%
Trimethyl orthopropionate
CAS: 24823-81-2; MDL No: MFCD00043305
UN No: UN3272; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2C(OCH3)3; Molecular Formula: C6H14O3; MW: 134.17
Boiling Point: 121-122°; Flash point: 19°C (66°F)
Density (g/mL): 0.944; Refractive Index: 1.398
- Refractive index: 1.398
- Liquid
Formula:
C₆H₁₄O₃ Boiling Pt: 121…122 °C Density: 0.944 g/mL Flash Pt: 19 °C (66 °F) |
MDL Number:
MFCD00043305 CAS Number: 24823-81-2 UN: 3272 ADR: 3,II |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...