1-Chlorophthalazine ≥97%

Supplier: BeanTown Chemical

221315-5G
BT221315-5GEA 192 USD
BT221315-5G
1-Chlorophthalazine ≥97%
1-Chlorophthalazine

CAS: 5784-45-2; MDL No: MFCD00024141 Solid; Molecular Formula: C8H5N2Cl; MW: 164.59


  • Solid

Formula: C₈H₅ClN₂
MDL Number: MFCD00024141
CAS Number: 5784-45-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR