Pyrocatechol ≥99%, Technical Grade
Supplier: BeanTown Chemical
CAS: 120-80-9; EC No: 204-427-5; MDL No: MFCD00002188; RTECS: UX1050000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C6H6O2; MW: 110.11
Melting Point: 100-103°; Boiling Point: 245°; Flash point: 127°C (261°F)
- RTECS: UX1050000
- Solid
Formula:
C₆H₆O₂ Boiling Pt: 245 °C Melting Pt: 100…103 °C Flash Pt: 127 °C (261 °F) |
MDL Number:
MFCD00002188 CAS Number: 120-80-9 EINECS: 204-427-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...